Anti-NMNAT1 Mouse Polyclonal Antibody

H00064802-B01P
10565-754EA 420.99 CAD
10565-754
Anti-NMNAT1 Mouse Polyclonal Antibody
Type: Primary
Antigen: NMNAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR