Anti-FNDC4 Rabbit Polyclonal Antibody

Supplier: Abnova
H00064838-D01
10586-478EA 564 CAD
10586-478
Anti-FNDC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FNDC4 (fibronectin type III domain containing 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR