Anti-FBXL17 Mouse Polyclonal Antibody

H00064839-B01P
10556-396EA 420.99 CAD
10556-396
Anti-FBXL17 Mouse Polyclonal Antibody
Type: Primary
Antigen: FBXL17 (F-box and leucine-rich repeat protein 17)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR