Anti-AIDA Mouse Monoclonal Antibody [clone: 2E6-1C11]

Supplier: Abnova
H00064853-M01
10586-182EA 502.5 CAD
10586-182
Anti-AIDA Mouse Monoclonal Antibody [clone: 2E6-1C11]
Antibodies
Type: Primary
Antigen: AIDA
Clonality: Monoclonal
Clone: 2E6-1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR