Anti-CENPH Mouse Polyclonal Antibody

Supplier: Abnova
H00064946-B02P
10553-194EA 502.5 CAD
10553-194
Anti-CENPH Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPH
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR