Anti-CENPH Rabbit Polyclonal Antibody

Supplier: Abnova
H00064946-D01P
10581-482EA 564 CAD
10581-482
Anti-CENPH Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPH
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR