Anti-ACD Mouse Monoclonal Antibody [clone: 1C11-1A7]

Supplier: Abnova
H00065057-M01
10576-064EA 502.5 CAD
10576-064
Anti-ACD Mouse Monoclonal Antibody [clone: 1C11-1A7]
Antibodies
Type: Primary
Antigen: ACD
Clonality: Monoclonal
Clone: 1C11-1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR