Anti-UPF3A Mouse Monoclonal Antibody [clone: 200]

Supplier: Abnova
H00065110-M06
10605-416EA 502.5 CAD
10605-416
Anti-UPF3A Mouse Monoclonal Antibody [clone: 200]
Antibodies
Type: Primary
Antigen: UPF3A
Clonality: Monoclonal
Clone: 200
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR