Anti-WNK2 Mouse Monoclonal Antibody [clone: 2A2]

Supplier: Abnova
H00065268-M02
10598-156EA 502.5 CAD
10598-156
Anti-WNK2 Mouse Monoclonal Antibody [clone: 2A2]
Antibodies
Type: Primary
Antigen: WNK2
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR