Anti-AACS Mouse Polyclonal Antibody

H00065985-B01
10509-976EA 408.81 CAD
10509-976
Anti-AACS Mouse Polyclonal Antibody
Type: Primary
Antigen: AACS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR