Anti-CYP4F12 Mouse Polyclonal Antibody

H00066002-B01P
10554-668EA 420.99 CAD
10554-668
Anti-CYP4F12 Mouse Polyclonal Antibody
Type: Primary
Antigen: CYP4F12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR