Anti-BOLL Mouse Monoclonal Antibody [clone: 1A7]

Supplier: Abnova
H00066037-M09
10578-028EA 502.5 CAD
10578-028
Anti-BOLL Mouse Monoclonal Antibody [clone: 1A7]
Antibodies
Type: Primary
Antigen: BOLL
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR