Anti-SLX1B Mouse Polyclonal Antibody

H00079008-B01P
10557-498EA 420.99 CAD
10557-498
Anti-SLX1B Mouse Polyclonal Antibody
Type: Primary
Antigen: SLX1B (SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR