Anti-CAMKV Mouse Monoclonal Antibody [clone: 2F3-1A2]

Supplier: Abnova
H00079012-M01
10593-378EA 502.5 CAD
10593-378
Anti-CAMKV Mouse Monoclonal Antibody [clone: 2F3-1A2]
Antibodies
Type: Primary
Antigen: CAMKV
Clonality: Monoclonal
Clone: 2F3-1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR