Anti-AHNAK Mouse Monoclonal Antibody [clone: 3G7]

Supplier: Abnova
H00079026-M01
10576-442EA 502.5 CAD
10576-442
Anti-AHNAK Mouse Monoclonal Antibody [clone: 3G7]
Antibodies
Type: Primary
Antigen: AHNAK
Clonality: Monoclonal
Clone: 3G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR