Anti-DDX54 Mouse Monoclonal Antibody [clone: 5B3]

Supplier: Abnova
H00079039-M03
10583-468EA 502.5 CAD
10583-468
Anti-DDX54 Mouse Monoclonal Antibody [clone: 5B3]
Antibodies
Type: Primary
Antigen: DDX54
Clonality: Monoclonal
Clone: 5B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR