Anti-RNF26 Mouse Monoclonal Antibody [clone: 5B9]

Supplier: Abnova
H00079102-M01
10599-988EA 502.5 CAD
10599-988
Anti-RNF26 Mouse Monoclonal Antibody [clone: 5B9]
Antibodies
Type: Primary
Antigen: RNF26
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR