Anti-NDUFAF5 Mouse Monoclonal Antibody [clone: 4A9]

Supplier: Abnova
H00079133-M01
10578-312EA 502.5 CAD
10578-312
Anti-NDUFAF5 Mouse Monoclonal Antibody [clone: 4A9]
Antibodies
Type: Primary
Antigen: NDUFAF5 (NADH dehydrogenase (ubiquinone) complex I, assembly factor 5)
Clonality: Monoclonal
Clone: 4A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR