Anti-NEIL1 Mouse Polyclonal Antibody

Supplier: Abnova
H00079661-B01
10565-502EA 502.5 CAD
10565-502
Anti-NEIL1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NEIL1 (nei endonuclease VIII-like 1 (E. coli))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR