Anti-SAP30L Mouse Polyclonal Antibody

H00079685-B01
10570-626EA 408.81 CAD
10570-626
Anti-SAP30L Mouse Polyclonal Antibody
Type: Primary
Antigen: SAP30L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR