Anti-GAL3ST4 Mouse Monoclonal Antibody [clone: 40000]

Supplier: Abnova
H00079690-M01
10586-992EA 502.5 CAD
10586-992
Anti-GAL3ST4 Mouse Monoclonal Antibody [clone: 40000]
Antibodies
Type: Primary
Antigen: GAL3ST4 (galactose-3-O-sulfotransferase 4)
Clonality: Monoclonal
Clone: 40000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR