Anti-GALNT12 Mouse Polyclonal Antibody

Supplier: Abnova
H00079695-B02
10557-280EA 502.5 CAD
10557-280
Anti-GALNT12 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GALNT12 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 (GalNAc-T12))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR