Anti-PRKRIP1 Mouse Monoclonal Antibody [clone: 3B11]

Supplier: Abnova
H00079706-M02
10597-586EA 502.5 CAD
10597-586
Anti-PRKRIP1 Mouse Monoclonal Antibody [clone: 3B11]
Antibodies
Type: Primary
Antigen: PRKRIP1
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR