Anti-LIN28 Rabbit Polyclonal Antibody

Supplier: Abnova
H00079727-D01
10591-830EA 564 CAD
10591-830
Anti-LIN28 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LIN28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR