Anti-CARF Mouse Polyclonal Antibody

Supplier: Abnova
H00079800-A01
10510-796EA 364.5 CAD
10510-796
Anti-CARF Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CARF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR