Anti-GSTCD Mouse Polyclonal Antibody

Supplier: Abnova
H00079807-B01
10560-968EA 502.5 CAD
10560-968
Anti-GSTCD Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GSTCD (glutathione S-transferase, C-terminal domain containing)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR