Anti-TLE6 Mouse Polyclonal Antibody

H00079816-B01
10572-442EA 392.79 CAD
10572-442
Anti-TLE6 Mouse Polyclonal Antibody
Type: Primary
Antigen: TLE6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR