Anti-TLE6 Mouse Monoclonal Antibody [clone: 20000]

Supplier: Abnova
H00079816-M01
10586-204EA 502.5 CAD
10586-204
Anti-TLE6 Mouse Monoclonal Antibody [clone: 20000]
Antibodies
Type: Primary
Antigen: TLE6
Clonality: Monoclonal
Clone: 20000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR