Anti-LONRF3 Mouse Monoclonal Antibody [clone: 1D5]

Supplier: Abnova
H00079836-M01
10599-892EA 502.5 CAD
10599-892
Anti-LONRF3 Mouse Monoclonal Antibody [clone: 1D5]
Antibodies
Type: Primary
Antigen: LONRF3
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR