Anti-THNSL1 (threonine Synthase-like 1 (S. cerevisiae)) Mouse Monoclonal Antibody [clone: 6B1]

Supplier: Abnova
H00079896-M08
10603-754EA 502.5 CAD
10603-754
Anti-THNSL1 (threonine Synthase-like 1 (S. cerevisiae)) Mouse Monoclonal Antibody [clone: 6B1]
Antibodies
Type: Primary
Antigen: THNSL1 (threonine Synthase-like 1 (S. cerevisiae))
Clonality: Monoclonal
Clone: 6B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR