Anti-SPEF2 Mouse Monoclonal Antibody [clone: 4B10]

Supplier: Abnova
H00079925-M01
10556-784EA 502.5 CAD
10556-784
Anti-SPEF2 Mouse Monoclonal Antibody [clone: 4B10]
Antibodies
Type: Primary
Antigen: SPEF2 (sperm flagellar 2)
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR