Anti-PLEKHS1 Mouse Polyclonal Antibody

Supplier: Abnova
H00079949-B01
10551-794EA 502.5 CAD
10551-794
Anti-PLEKHS1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLEKHS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR