Anti-PGAP1 Mouse Monoclonal Antibody [clone: 5G6]

Supplier: Abnova
H00080055-M01
10596-772EA 502.5 CAD
10596-772
Anti-PGAP1 Mouse Monoclonal Antibody [clone: 5G6]
Antibodies
Type: Primary
Antigen: PGAP1
Clonality: Monoclonal
Clone: 5G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR