Anti-DBF4B Mouse Polyclonal Antibody

Supplier: Abnova
H00080174-B01P
10554-730EA 502.5 CAD
10554-730
Anti-DBF4B Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DBF4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR