Anti-CXORF21 Mouse Polyclonal Antibody

H00080231-B01
10554-380EA 420.99 CAD
10554-380
Anti-CXORF21 Mouse Polyclonal Antibody
Type: Primary
Antigen: CXORF21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR