Anti-GRPEL1 Mouse Polyclonal Antibody

H00080273-B01P
10557-924EA 420.99 CAD
10557-924
Anti-GRPEL1 Mouse Polyclonal Antibody
Type: Primary
Antigen: GRPEL1 (GrpE-like 1, mitochondrial (E. coli))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR