Anti-EPC1 Mouse Monoclonal Antibody [clone: 4F7]

H00080314-M01A
10624-256EA 420.99 CAD
10624-256
Anti-EPC1 Mouse Monoclonal Antibody [clone: 4F7]
Type: Primary
Antigen: EPC1
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR