Anti-PUS1 Mouse Monoclonal Antibody [clone: 3A3]

Supplier: Abnova
H00080324-M01
10598-772EA 502.5 CAD
10598-772
Anti-PUS1 Mouse Monoclonal Antibody [clone: 3A3]
Antibodies
Type: Primary
Antigen: PUS1
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR