Anti-DNAJC5 Mouse Polyclonal Antibody

H00080331-B01P
10555-264EA 420.99 CAD
10555-264
Anti-DNAJC5 Mouse Polyclonal Antibody
Type: Primary
Antigen: DNAJC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR