Anti-KCNIP4 Mouse Monoclonal Antibody [clone: 1A11]

Supplier: Abnova
H00080333-M01
10589-680EA 502.5 CAD
10589-680
Anti-KCNIP4 Mouse Monoclonal Antibody [clone: 1A11]
Antibodies
Type: Primary
Antigen: KCNIP4
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR