Anti-TSEN2 Mouse Polyclonal Antibody

H00080746-B01P
10573-090EA 420.99 CAD
10573-090
Anti-TSEN2 Mouse Polyclonal Antibody
Type: Primary
Antigen: TSEN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR