Anti-ZNF34 Mouse Monoclonal Antibody [clone: 1B11]

Supplier: Abnova
H00080778-M03
10606-484EA 502.5 CAD
10606-484
Anti-ZNF34 Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
Type: Primary
Antigen: ZNF34
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR