Anti-APOL3 Mouse Monoclonal Antibody [clone: 8F5]

Supplier: Abnova
H00080833-M02
10577-140EA 502.5 CAD
10577-140
Anti-APOL3 Mouse Monoclonal Antibody [clone: 8F5]
Antibodies
Type: Primary
Antigen: APOL3
Clonality: Monoclonal
Clone: 8F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR