Anti-SLC2A10 Mouse Monoclonal Antibody [clone: 1E12]

Supplier: Abnova
H00081031-M02
10601-626EA 502.5 CAD
10601-626
Anti-SLC2A10 Mouse Monoclonal Antibody [clone: 1E12]
Antibodies
Type: Primary
Antigen: SLC2A10
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR