Anti-COLEC12 Mouse Polyclonal Antibody

Supplier: Abnova
H00081035-B01
10559-950EA 502.5 CAD
10559-950
Anti-COLEC12 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COLEC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR