Anti-FAM117A Rabbit Polyclonal Antibody

Supplier: Abnova
H00081558-D01P
10585-616EA 564 CAD
10585-616
Anti-FAM117A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FAM117A (family with sequence similarity 117, member A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR