Anti-PLA2G12A Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Abnova
H00081579-M01
10596-594EA 502.5 CAD
10596-594
Anti-PLA2G12A Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Type: Primary
Antigen: PLA2G12A
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR