Anti-ITM2C Mouse Polyclonal Antibody

H00081618-B01P
10562-040EA 420.99 CAD
10562-040
Anti-ITM2C Mouse Polyclonal Antibody
Type: Primary
Antigen: ITM2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR