Anti-TSSK3 Mouse Monoclonal Antibody [clone: 3F9]

Supplier: Abnova
H00081629-M03
10602-694EA 502.5 CAD
10602-694
Anti-TSSK3 Mouse Monoclonal Antibody [clone: 3F9]
Antibodies
Type: Primary
Antigen: TSSK3
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR