Anti-MAP1LC3B Mouse Monoclonal Antibody [clone: 4G7]

Supplier: Abnova
H00081631-M02
10592-540EA 502.5 CAD
10592-540
Anti-MAP1LC3B Mouse Monoclonal Antibody [clone: 4G7]
Antibodies
Type: Primary
Antigen: MAP1LC3B
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR